2,4,6-Tribromobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

108102-5G 108102-1G 108102-500MG
77653-194EA 2077 USD
77653-194 77640-330 77648-326
2,4,6-Tribromobenzonitrile ≥97%
2,4,6-Tribromobenzonitrile

Precaución: For Research Use Only

Formula: 339.809 g/mol
MW: C₇H₂Br₃N
Storage Temperature: Ambient
MDL Number: MFCD06797959
CAS Number: 35851-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR