2,3-Dichloro-4-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

107997-5G 107997-25G 107997-1G
77633-910EA 194 USD
77633-910 77657-180 77640-328
2,3-Dichloro-4-methoxybenzaldehyde ≥95%
2,3-Dichloro-4-methoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22493491
CAS Number: 41827-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR