1-(5-Bromo-2,3,4-trifluorophenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

107949-5G 107949-1G 107949-25G 107949-500MG
77631-010EA 1484.23 USD
77631-010 77636-316 77653-192 77644-320
1-(5-Bromo-2,3,4-trifluorophenyl)-2-fluoroethanone ≥95%
1-(5-Bromo-2,3,4-trifluorophenyl)-2-fluoroethanone

Precaución: For Research Use Only

Formula: 271.006 g/mol
MW: C₈H₃BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD34779664
CAS Number: 2918910-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR