N-methoxy-N,3,5-trimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

107589-5G 107589-25G 107589-1G 107589-500MG
77631-416EA 872 USD
77631-416 77653-190 77640-326 77648-322
N-methoxy-N,3,5-trimethylbenzamide ≥97%
N-methoxy-N,3,5-trimethylbenzamide

Precaución: For Research Use Only

Formula: C₁₁H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD14707531
CAS Number: 252199-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR