4-Bromo-2-chloro-1-cyclobutoxybenzene ≥97%

Supplier: AOBChem USA

Warning

10700-5G 10700-1G 10700-500MG
77653-186EA 1992 USD
77653-186 77644-316 77648-320
4-Bromo-2-chloro-1-cyclobutoxybenzene ≥97%
4-Bromo-2-chloro-1-cyclobutoxybenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₀BrClO
Storage Temperature: Ambient
MDL Number: MFCD30099130
CAS Number: 1683546-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR