3-Bromo-2-chloro-4-fluorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

106726-1G 106726-5G 106726-500MG
77633-908EA 819 USD
77633-908 77657-178 77648-318
3-Bromo-2-chloro-4-fluorobenzonitrile ≥97%
3-Bromo-2-chloro-4-fluorobenzonitrile

Precaución: For Research Use Only

Formula: 234.453 g/mol
MW: C₇H₂BrClFN
Storage Temperature: Ambient
MDL Number: MFCD33482350
CAS Number: 2091679-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR