5-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

106687-5G 106687-25G 106687-1G 106687-500MG
77631-412EA 944 USD
77631-412 77657-176 77640-322 77648-316
5-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile ≥95%
5-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile

Precaución: For Research Use Only

Formula: C₈H₂BrF₄N
MW: 268.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30829166
CAS Number: 2021202-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR