4-Isopropyl-3-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

106426-5G 106426-1G 106426-500MG
77631-008EA 1560.45 USD
77631-008 77640-320 77648-314
4-Isopropyl-3-methoxybenzoic acid ≥95%
4-Isopropyl-3-methoxybenzoic acid

Precaución: For Research Use Only

Formula: 194.227 g/mol
MW: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD21602289
CAS Number: 1361003-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR