1,2,3-Tribromo-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

106256-5G 106256-25G
77633-906EA 154 USD
77633-906 77657-174
1,2,3-Tribromo-5-methylbenzene ≥95%
1,2,3-Tribromo-5-methylbenzene

Precaución: For Research Use Only

Formula: 328.827 g/mol
MW: C₇H₅Br₃
Storage Temperature: Ambient
MDL Number: MFCD00075798
CAS Number: 73557-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR