2'-Bromo-4'-methoxyacetophenone ≥95%

Supplier: AOBChem USA

Warning

105747-5G 105747-25G
77633-904EA 66 USD
77633-904 77657-172
2'-Bromo-4'-methoxyacetophenone ≥95%
2'-Bromo-4'-methoxyacetophenone

Precaución: For Research Use Only

Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD00465443
CAS Number: 89691-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR