1-Bromo-5-ethyl-2,3-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

104896-5G 104896-1G 104896-500MG
77653-178EA 2023 USD
77653-178 77640-314 77648-306
1-Bromo-5-ethyl-2,3-dimethoxybenzene ≥97%
1-Bromo-5-ethyl-2,3-dimethoxybenzene

Precaución: For Research Use Only

Formula: 245.113 g/mol
MW: C₁₀H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD34820691
CAS Number: 849669-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR