2-Methoxy-5-propylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

104545-5G 104545-1G 104545-500MG
77657-168EA 2083 USD
77657-168 77640-312 77644-310
2-Methoxy-5-propylbenzoic acid ≥95%
2-Methoxy-5-propylbenzoic acid

Precaución: For Research Use Only

Formula: 194.227 g/mol
MW: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD20644539
CAS Number: 130880-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR