(2-Methoxy-5-propylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

104542-5G 104542-1G 104542-25G 104542-500MG
77631-406EA 1441 USD
77631-406 77636-308 77657-166 77648-304
(2-Methoxy-5-propylphenyl)boronic acid ≥95%
(2-Methoxy-5-propylphenyl)boronic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD18384171
CAS Number: 1261181-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR