1-(Benzyloxy)-2-bromo-4-propylbenzene ≥97%

Supplier: AOBChem USA

Warning

104534-5G 104534-1G 104534-25G 104534-500MG
77631-002EA 1484.23 USD
77631-002 77636-306 77653-174 77644-308
1-(Benzyloxy)-2-bromo-4-propylbenzene ≥97%
1-(Benzyloxy)-2-bromo-4-propylbenzene

Precaución: For Research Use Only

Formula: 305.21 g/mol
MW: C₁₆H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD34820602
CAS Number: 2918953-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR