4-(4-(Trimethylsilyl)phenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

104313-5G 104313-25G 104313-1G 104313-500MG
77631-000EA 1688.17 USD
77631-000 77635-394 77640-310 77648-302
4-(4-(Trimethylsilyl)phenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(4-(Trimethylsilyl)phenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 250.409 g/mol
MW: C₁₄H₂₂O₂Si
Storage Temperature: Ambient
MDL Number: MFCD34778065
CAS Number: 2237250-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR