4-(Naphthalen-2-yl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

104297-5G 104297-1G 104297-25G 104297-500MG
77631-404EA 1381 USD
77631-404 77636-304 77653-172 77644-306
4-(Naphthalen-2-yl)tetrahydro-2H-pyran-4-ol ≥95%
4-(Naphthalen-2-yl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: C₁₅H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD19461670
CAS Number: 1282493-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR