4-(Naphthalen-1-yl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

104296-5G 104296-1G 104296-25G 104296-500MG
77631-402EA 1381 USD
77631-402 77636-302 77657-164 77648-300
4-(Naphthalen-1-yl)tetrahydro-2H-pyran-4-ol ≥95%
4-(Naphthalen-1-yl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: C₁₅H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD18549616
CAS Number: 1280222-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR