4-(3,4-Difluoro-2-methylphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

104255-5G 104255-25G 104255-1G 104255-500MG
77630-998EA 1484.23 USD
77630-998 77635-392 77636-300 77648-298
4-(3,4-Difluoro-2-methylphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3,4-Difluoro-2-methylphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 228.235 g/mol
MW: C₁₂H₁₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34778016
CAS Number: 2918883-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR