1-(3,4-Difluoro-2-methylphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

104254-5G 104254-25G 104254-1G 104254-500MG
77630-996EA 1720.1 USD
77630-996 77653-170 77640-308 77648-296
1-(3,4-Difluoro-2-methylphenyl)cyclohexanol ≥95%
1-(3,4-Difluoro-2-methylphenyl)cyclohexanol

Precaución: For Research Use Only

Formula: 226.262 g/mol
MW: C₁₃H₁₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD34778015
CAS Number: 2918920-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR