4-(3,4-Dichlorophenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

104070-5G 104070-1G 104070-500MG
77630-988EA 1553.24 USD
77630-988 77640-300 77644-298
4-(3,4-Dichlorophenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3,4-Dichlorophenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 247.118 g/mol
MW: C₁₁H₁₂Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18296161
CAS Number: 1343942-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR