4-(2,5-Dimethylphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

104067-5G 104067-25G 104067-1G 104067-500MG
77631-400EA 1206 USD
77631-400 77653-166 77640-298 77648-292
4-(2,5-Dimethylphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(2,5-Dimethylphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 206.281 g/mol
MW: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD18296194
CAS Number: 1340076-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR