4-(4-Fluoro-2-methoxyphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

104061-5G 104061-25G 104061-1G 104061-500MG
77630-984EA 1615.04 USD
77630-984 77635-386 77640-294 77644-292
4-(4-Fluoro-2-methoxyphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(4-Fluoro-2-methoxyphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 226.244 g/mol
MW: C₁₂H₁₅FO₃
Storage Temperature: Ambient
MDL Number: MFCD31694061
CAS Number: 2254571-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR