4-(3,4-Dimethylphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

104059-5G 104059-1G 104059-500MG
77630-982EA 1615.04 USD
77630-982 77640-290 77644-288
4-(3,4-Dimethylphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3,4-Dimethylphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 206.281 g/mol
MW: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD18296150
CAS Number: 1340153-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR