1-(2-Fluoro-3-(trifluoromethyl)phenyl)cyclobutanol ≥95%
Supplier: AOBChem USA
|
Warning
|
104027-5G
104027-1G
104027-25G
104027-500MG
77630-980EA
1553.24
USD
77630-980
77636-294
77657-160
77644-286
1-(2-Fluoro-3-(trifluoromethyl)phenyl)cyclobutanol ≥95%
1-(2-Fluoro-3-(trifluoromethyl)phenyl)cyclobutanol
Precaución: For Research Use Only
Formula:
C₁₁H₁₀F₄O Storage Temperature: Ambient |
MDL Number:
MFCD34777841 CAS Number: 2918907-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...