1-(4-Bromo-1-fluoronaphthalen-2-yl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

103419-5G 103419-25G 103419-1G 103419-500MG
77630-978EA 1484.23 USD
77630-978 77657-158 77640-288 77644-284
1-(4-Bromo-1-fluoronaphthalen-2-yl)cyclobutanol ≥95%
1-(4-Bromo-1-fluoronaphthalen-2-yl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₄H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD34777598
CAS Number: 2918905-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR