1-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

103404-5G 103404-1G 103404-25G 103404-500MG
77630-976EA 1553.24 USD
77630-976 77636-292 77653-160 77648-290
1-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)cyclobutanol ≥95%
1-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 268.635 g/mol
MW: C₁₁H₉ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD34777585
CAS Number: 2918904-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR