1-(2,4-Difluoro-3-methoxyphenyl)cyclobutanol ≥97%

Supplier: AOBChem USA

Warning

103401-5G 103401-1G 103401-25G 103401-500MG
77630-974EA 1553.24 USD
77630-974 77636-290 77653-158 77648-288
1-(2,4-Difluoro-3-methoxyphenyl)cyclobutanol ≥97%
1-(2,4-Difluoro-3-methoxyphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 214.209 g/mol
MW: C₁₁H₁₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34777582
CAS Number: 2918839-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR