1-(2-Fluoro-5-iodophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

103377-5G 103377-1G 103377-25G 103377-500MG
77630-972EA 1553.24 USD
77630-972 77636-288 77653-156 77644-282
1-(2-Fluoro-5-iodophenyl)cyclobutanol ≥95%
1-(2-Fluoro-5-iodophenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 292.089 g/mol
MW: C₁₀H₁₀FIO
Storage Temperature: Ambient
MDL Number: MFCD34777559
CAS Number: 2918877-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR