1,2-Dibromo-3-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

103294-5G 103294-25G
77633-902EA 171 USD
77633-902 77657-156
1,2-Dibromo-3-fluoro-4-methoxybenzene ≥97%
1,2-Dibromo-3-fluoro-4-methoxybenzene

Precaución: For Research Use Only

Formula: 283.92 g/mol
MW: C₇H₅Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD22205768
CAS Number: 1375069-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR