1-(3-Chloro-2-fluorophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

102707-5G 102707-1G 102707-25G 102707-500MG
77630-970EA 1777.78 USD
77630-970 77636-286 77657-154 77648-286
1-(3-Chloro-2-fluorophenyl)cyclohexanol ≥95%
1-(3-Chloro-2-fluorophenyl)cyclohexanol

Precaución: For Research Use Only

Formula: 228.69 g/mol
MW: C₁₂H₁₄ClFO
Storage Temperature: Ambient
MDL Number: MFCD34777284
CAS Number: 2918913-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR