1-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

102665-5G 102665-500MG 102665-25G 102665-1G
77630-966EA 1484.23 USD
77630-966 77652-274 77653-152 77640-284
1-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)cyclohexanol ≥97%
1-(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)cyclohexanol

Precaución: For Research Use Only

Formula: 296.688 g/mol
MW: C₁₃H₁₃ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD34777250
CAS Number: 2901109-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR