3-Iodo-2,5-dimethoxypyridine ≥95%

Supplier: AOBChem USA

Warning

102313-5G 102313-25G 102313-1G 102313-500MG
77630-964EA 1484.23 USD
77630-964 77635-384 77636-284 77644-280
3-Iodo-2,5-dimethoxypyridine ≥95%
3-Iodo-2,5-dimethoxypyridine

Precaución: For Research Use Only

Formula: C₇H₈INO₂
Storage Temperature: Ambient
MDL Number: MFCD34776964
CAS Number: 2918950-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR