3-Fluoro-2,4-bis(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

102264-5G 102264-1G 102264-25G 102264-500MG
77630-960EA 1484.23 USD
77630-960 77636-282 77653-148 77648-280
3-Fluoro-2,4-bis(methylthio)pyridine ≥95%
3-Fluoro-2,4-bis(methylthio)pyridine

Precaución: For Research Use Only

Formula: C₇H₈FNS₂
MW: 189.274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34776919
CAS Number: 902518-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR