4-Benzyloxy-2-piperidine-1-yl-pyrimidine-5-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

10211-5G 10211-500MG 10211-25G 10211-1G
77631-394EA 648 USD
77631-394 77652-272 77657-150 77640-278
4-Benzyloxy-2-piperidine-1-yl-pyrimidine-5-boronic acid pinacol ester ≥97%
4-Benzyloxy-2-piperidine-1-yl-pyrimidine-5-boronic acid pinacol ester

Precaución: For Research Use Only

Formula: 395.303 g/mol
MW: C₂₂H₃₀BN₃O₃
Storage Temperature: Ambient
MDL Number: MFCD09839110
CAS Number: 1451391-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR