2-Bromo-4,6-dichlorobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

101884-5G 101884-1G 101884-500MG
77653-144EA 2402 USD
77653-144 77640-276 77648-278
2-Bromo-4,6-dichlorobenzonitrile ≥95%
2-Bromo-4,6-dichlorobenzonitrile

Precaución: For Research Use Only

Formula: C₇H₂BrCl₂N
Storage Temperature: Ambient
MDL Number: MFCD31668158
CAS Number: 99835-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR