4-Hydroxy-2,5-dimethoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

101418-1G 101418-5G 101418-500MG
77636-278EA 556 USD
77636-278 77653-142 77648-274
4-Hydroxy-2,5-dimethoxybenzaldehyde ≥95%
4-Hydroxy-2,5-dimethoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00597677
CAS Number: 80749-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR