Methyl 6-bromo-2-fluoro-3-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

101218-5G 101218-1G 101218-500MG 101218-25G
77633-900EA 414 USD
77633-900 77636-276 77652-270 77657-146
Methyl 6-bromo-2-fluoro-3-(trifluoromethyl)benzoate ≥95%
Methyl 6-bromo-2-fluoro-3-(trifluoromethyl)benzoate

Precaución: For Research Use Only

Formula: C₉H₅BrF₄O₂
MW: 301.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28134146
CAS Number: 1352718-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR