(3-Isopropyl-4-methylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

101189-1G 101189-5G 101189-500MG
77636-274EA 535 USD
77636-274 77657-144 77644-272
(3-Isopropyl-4-methylphenyl)boronic acid ≥95%
(3-Isopropyl-4-methylphenyl)boronic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₅BO₂
Storage Temperature: Ambient
MDL Number: MFCD34776459
CAS Number: 2412794-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR