2-Bromo-7-ethoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

100954-5G 100954-25G 100954-1G 100954-500MG
77631-392EA 904 USD
77631-392 77657-142 77640-274 77648-272
2-Bromo-7-ethoxynaphthalene ≥97%
2-Bromo-7-ethoxynaphthalene

Precaución: For Research Use Only

Formula: 251.119 g/mol
MW: C₁₂H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD17012562
CAS Number: 627527-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR