5-Methoxynaphthalen-1-ol ≥97%

Supplier: AOBChem USA

Warning

100873-5G 100873-25G 100873-1G 100873-500MG
77631-390EA 713 USD
77631-390 77657-138 77640-270 77648-270
5-Methoxynaphthalen-1-ol ≥97%
5-Methoxynaphthalen-1-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00512042
CAS Number: 3588-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR