8-Bromo-1-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

100805-5G 100805-1G 100805-25G 100805-500MG
77631-388EA 1081 USD
77631-388 77636-270 77657-136 77648-268
8-Bromo-1-naphthaldehyde ≥97%
8-Bromo-1-naphthaldehyde

Precaución: For Research Use Only

Formula: C₁₁H₇BrO
Storage Temperature: Ambient
MDL Number: MFCD09909583
CAS Number: 85864-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR