3-Chloro-2-fluorobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Chloro-2-fluoro-3-(trifluoromethyl)benzene

100403-5G 100403-25G
77633-898EA 67 USD
77633-898 77653-138
3-Chloro-2-fluorobenzotrifluoride ≥97%
3-Chloro-2-fluorobenzotrifluoride

Precaución: For Research Use Only

Formula: C₇H₃ClF₄
MW: 198.5500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100532
CAS Number: 1099597-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR