4-Formyl-2-methoxy-benzonitrile 95%

Supplier: Ambeed
Danger

A593708-5G A593708-1G
77673-592EA 239 USD
77673-592 77884-429
4-Formyl-2-methoxy-benzonitrile 95%
4-Formyl-2-methoxy-benzonitrile

Precaución: for Research Use Only

Formula: C₉H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD16295254
CAS Number: 21962-49-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR