UK-78282 hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: UK-78282 monohydrochloride, 4-((Benzhydryloxy)methyl)-1-(3-(4-methoxyphenyl)propyl)piperidine hydrochloride

A570533-10MG A570533-5MG A570533-100MG A570533-250MG A570533-50MG
77673-526EA 80 USD
77673-526 77673-528 77884-392 77884-393 77884-394
UK-78282 hydrochloride 98%
UK-78282 hydrochloride

Precaución: for Research Use Only

Formula: C₂₉H₃₆ClNO₂
MW: 466.05 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16875438
CAS Number: 136647-02-4
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR