3-(Methoxymethyl)cyclobutanone 97%

Supplier: Ambeed
Danger

A897385-1G A897385-250MG
77623-120EA 219.39 USD
77623-120 77623-122
3-(Methoxymethyl)cyclobutanone 97%
3-(Methoxymethyl)cyclobutanone

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22381625
CAS Number: 1068160-23-5
UN: 1224
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR