2-Bromo-5,6-dihydro-4H-cyclopenta[d]thiazole 98%

Supplier: Ambeed
Warning

A820922-100MG A820922-1G A820922-250MG
77674-210EA 160 USD
77674-210 77674-212 77674-214
2-Bromo-5,6-dihydro-4H-cyclopenta[d]thiazole 98%
2-Bromo-5,6-dihydro-4H-cyclopenta[d]thiazole

Precaución: for Research Use Only

Formula: C₆H₆BrNS
MW: 204.09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD20921667
CAS Number: 1080642-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR