4-Fmoc-1-piperazineacetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperazin-1-yl)acetic acid, Fmoc-(4-carboxymethyl)piperazine

A542087-1G A542087-5G
77673-450EA 50 USD
77673-450 77673-452
4-Fmoc-1-piperazineacetic acid 95%
4-Fmoc-1-piperazineacetic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₂N₂O₄
MW: 366.41 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270202
CAS Number: 180576-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR