Dabsyl chloride 98%

Supplier: Ambeed
Danger

A272423-1G A272423-5G A272423-10G A272423-25G
77626-412EA 81.37 USD
77626-412 77626-414 77879-502 77879-503
Dabsyl chloride 98%
Dabsyl chloride

Precaución: for Research Use Only

Formula: C₁₄H₁₄ClN₃O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00007444
CAS Number: 56512-49-3
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR