YK-029A 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-((2-(Dimethylamino)ethyl)(methyl)amino)-4-methoxy-5-((4-(8-methylimidazo[1,2-a]pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)acrylamide

A2420741-100MG A2420741-10MG A2420741-250MG A2420741-50MG A2420741-25MG
77626-286EA 644.78 USD
77626-286 77626-288 77626-290 77626-292 77877-805
YK-029A 98%
YK-029A

Precaución: for Research Use Only

Formula: C₂₇H₃₂N₈O₂
MW: 500.60 g/mol
Storage Temperature: Refrigerator
CAS Number: 2064269-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR