Cisd2 agonist 1 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-5-((4-fluorophenyl)carbamoyl)-4-methylthiophene-3-carboxylate

A2318419-25MG A2318419-50MG
77626-222EA 471.74 USD
77626-222 77626-224
Cisd2 agonist 1 98%
Cisd2 agonist 1

Precaución: for Research Use Only

Formula: C₁₄H₁₃FN₂O₃S
MW: 308.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD35068596
CAS Number: 2916371-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR